Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGCATCACCACCAGGTTCACGC[C/G]GAGAAGGAGCTGGAGGAGAGCCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171650 MIM: 600811 MIM: 602423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACP2 - acid phosphatase 2, lysosomal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302489.1 | 112 | Intron | NP_001289418.1 | |||
NM_001302490.1 | 112 | Missense Mutation | CGC,GGC | R,G 19 | NP_001289419.1 | |
NM_001302491.1 | 112 | Missense Mutation | CGC,GGC | R,G 19 | NP_001289420.1 | |
NM_001302492.1 | 112 | UTR 5 | NP_001289421.1 | |||
NM_001610.3 | 112 | Missense Mutation | CGC,GGC | R,G 19 | NP_001601.1 |
DDB2 - damage specific DNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |