Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCATTTTTTGTTTGCGTCCCCC[T/C]CCCGCGCCTCCTCTCCCTGCCGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609477 | ||||||||||||||||||||
Literature Links: |
SMCO4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SMCO4 - single-pass membrane protein with coiled-coil domains 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020179.2 | 1250 | Intron | NP_064564.1 | |||
XM_011542907.1 | 1250 | Intron | XP_011541209.1 | |||
XM_011542908.2 | 1250 | Intron | XP_011541210.1 | |||
XM_011542909.1 | 1250 | Intron | XP_011541211.1 | |||
XM_011542910.2 | 1250 | Missense Mutation | GAG,GGG | E,G 162 | XP_011541212.1 | |
XM_011542911.2 | 1250 | Missense Mutation | GAG,GGG | E,G 136 | XP_011541213.1 | |
XM_017018019.1 | 1250 | Missense Mutation | GAG,GGG | E,G 157 | XP_016873508.1 | |
XM_017018020.1 | 1250 | Intron | XP_016873509.1 |