Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGCTGGCTGTTGCCCCTGGACT[A/G]AGATTTCTTCCTCTTTGCTGGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610495 MIM: 164031 | ||||||||||||||||||||
Literature Links: |
IFFO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFFO1 - intermediate filament family orphan 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039670.2 | 2522 | Intron | NP_001034759.1 | |||
NM_001193457.1 | 2522 | Intron | NP_001180386.1 | |||
NM_080730.4 | 2522 | Intron | NP_542768.2 | |||
XM_006718975.3 | 2522 | Intron | XP_006719038.1 | |||
XM_006718976.3 | 2522 | Intron | XP_006719039.1 | |||
XM_006718977.3 | 2522 | Intron | XP_006719040.1 | |||
XM_006718981.3 | 2522 | Intron | XP_006719044.1 | |||
XM_011520947.2 | 2522 | Intron | XP_011519249.1 | |||
XM_011520948.2 | 2522 | Intron | XP_011519250.1 | |||
XM_011520949.2 | 2522 | Intron | XP_011519251.1 | |||
XM_011520952.2 | 2522 | Intron | XP_011519254.1 | |||
XM_017019169.1 | 2522 | Intron | XP_016874658.1 | |||
XM_017019170.1 | 2522 | Intron | XP_016874659.1 | |||
XM_017019171.1 | 2522 | Intron | XP_016874660.1 | |||
XM_017019172.1 | 2522 | Intron | XP_016874661.1 | |||
XM_017019173.1 | 2522 | Intron | XP_016874662.1 |
NOP2 - NOP2 nucleolar protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033714.2 | 2522 | Nonsense Mutation | CAG,TAG | Q,* 798 | NP_001028886.1 | |
NM_001258308.1 | 2522 | Nonsense Mutation | CAG,TAG | Q,* 802 | NP_001245237.1 | |
NM_001258309.1 | 2522 | Nonsense Mutation | CAG,TAG | Q,* 835 | NP_001245238.1 | |
NM_001258310.1 | 2522 | UTR 3 | NP_001245239.1 | |||
NM_006170.3 | 2522 | Nonsense Mutation | CAG,TAG | Q,* 798 | NP_006161.2 | |
XM_005253691.1 | 2522 | Nonsense Mutation | CAG,TAG | Q,* 802 | XP_005253748.1 |