Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTCCAGGAGACCCTGTGAAGC[C/T]GTCTCGGGGCCCGCTGGTGACCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601284 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACVRL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACVRL1 - activin A receptor like type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000020.2 | 223 | Missense Mutation | CCG,CTG | P,L 26 | NP_000011.2 | |
NM_001077401.1 | 223 | Missense Mutation | CCG,CTG | P,L 26 | NP_001070869.1 | |
XM_005269235.2 | 223 | Missense Mutation | CCG,CTG | P,L 26 | XP_005269292.1 | |
XM_011539008.1 | 223 | Missense Mutation | CCG,CTG | P,L 40 | XP_011537310.1 |