Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCTTTACACATATAGTTCCTTA[C/T]TGTAGAAGTAAAAAGAAAAAAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ACTR6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTR6 - ARP6 actin-related protein 6 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022496.4 | 1245 | Silent Mutation | TAC,TAT | Y,Y 160 | NP_071941.1 | |
XM_017019821.1 | 1245 | Silent Mutation | TAC,TAT | Y,Y 52 | XP_016875310.1 |
DEPDC4 - DEP domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319310.1 | 1245 | Intron | NP_001306239.1 | |||
NM_001319311.1 | 1245 | Intron | NP_001306240.1 | |||
NM_152317.3 | 1245 | Intron | NP_689530.1 | |||
XM_011537876.2 | 1245 | Intron | XP_011536178.1 | |||
XM_017018779.1 | 1245 | Intron | XP_016874268.1 | |||
XM_017018780.1 | 1245 | Intron | XP_016874269.1 | |||
XM_017018781.1 | 1245 | Intron | XP_016874270.1 | |||
XM_017018782.1 | 1245 | Intron | XP_016874271.1 | |||
XM_017018783.1 | 1245 | Intron | XP_016874272.1 | |||
XM_017018784.1 | 1245 | Intron | XP_016874273.1 | |||
XM_017018785.1 | 1245 | Intron | XP_016874274.1 |