Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACCCTGGTGCACTTCGTGGTC[A/G]CCTGGCTGGGCTTGTATATCTGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607617 | ||||||||||||||||||||
Literature Links: |
NUP107 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUP107 - nucleoporin 107 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC35E3 - solute carrier family 35 member E3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018656.2 | 390 | Missense Mutation | ACC,GCC | T,A 54 | NP_061126.2 | |
XM_005269006.3 | 390 | Missense Mutation | ACC,GCC | T,A 54 | XP_005269063.1 | |
XM_017019584.1 | 390 | Missense Mutation | ACC,GCC | T,A 54 | XP_016875073.1 |