Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTTCTTGGCTGGCTGGCTGCT[A/G]TTCTCAGACTTGGGGATGACCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610495 MIM: 164031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFFO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFFO1 - intermediate filament family orphan 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOP2 - NOP2 nucleolar protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033714.2 | 1972 | Silent Mutation | AAC,AAT | N,N 614 | NP_001028886.1 | |
NM_001258308.1 | 1972 | Silent Mutation | AAC,AAT | N,N 618 | NP_001245237.1 | |
NM_001258309.1 | 1972 | Silent Mutation | AAC,AAT | N,N 651 | NP_001245238.1 | |
NM_001258310.1 | 1972 | UTR 3 | NP_001245239.1 | |||
NM_006170.3 | 1972 | Silent Mutation | AAC,AAT | N,N 614 | NP_006161.2 | |
XM_005253691.1 | 1972 | Silent Mutation | AAC,AAT | N,N 618 | XP_005253748.1 |