Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACACAGCTGCCTCCCGGCACCGC[C/T]GCCACTCCTCGTCACTGTCCTCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142410 | ||||||||||||||||||||
Literature Links: |
C12orf43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C12orf43 - chromosome 12 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286191.1 | 408 | Missense Mutation | CAG,CGG | Q,R 190 | NP_001273120.1 | |
NM_001286192.1 | 408 | Missense Mutation | CAG,CGG | Q,R 160 | NP_001273121.1 | |
NM_001286195.1 | 408 | Missense Mutation | CAG,CGG | Q,R 149 | NP_001273124.1 | |
NM_001286196.1 | 408 | Missense Mutation | CAG,CGG | Q,R 148 | NP_001273125.1 | |
NM_001286197.1 | 408 | Missense Mutation | CAG,CGG | Q,R 127 | NP_001273126.1 | |
NM_001286198.1 | 408 | Missense Mutation | CAG,CGG | Q,R 118 | NP_001273127.1 | |
NM_022895.2 | 408 | Missense Mutation | CAG,CGG | Q,R 159 | NP_075046.1 | |
XM_017019829.1 | 408 | Intron | XP_016875318.1 |
HNF1A - HNF1 homeobox A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |