Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTCCAGTGGAGTAATTTAGCTT[C/T]CTGCAGAAAATGCTCCGAATATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601423 | ||||||||||||||||||||
Literature Links: |
GLT8D2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLT8D2 - glycosyltransferase 8 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316967.1 | 1943 | Missense Mutation | AAA,GAA | K,E 308 | NP_001303896.1 | |
NM_031302.4 | 1943 | Missense Mutation | AAA,GAA | K,E 308 | NP_112592.1 | |
XM_011538793.2 | 1943 | Missense Mutation | AAA,GAA | K,E 308 | XP_011537095.1 | |
XM_017019999.1 | 1943 | Missense Mutation | AAA,GAA | K,E 308 | XP_016875488.1 |
TDG - thymine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003211.4 | 1943 | Intron | NP_003202.3 | |||
XM_005269125.1 | 1943 | Intron | XP_005269182.1 | |||
XM_011538714.1 | 1943 | Intron | XP_011537016.1 | |||
XM_011538715.1 | 1943 | Intron | XP_011537017.1 |