Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCAAACCCCAGCCAGCCGTGCTC[A/G]AAAGCCTCCATTGGGTTATTTGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604482 | ||||||||||||||||||||
Literature Links: |
SIRT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIRT4 - sirtuin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012240.2 | 316 | Silent Mutation | TCA,TCG | S,S 25 | NP_036372.1 | |
XM_005253865.4 | 316 | Silent Mutation | TCA,TCG | S,S 25 | XP_005253922.1 | |
XM_006719308.2 | 316 | Silent Mutation | TCA,TCG | S,S 25 | XP_006719371.1 | |
XM_006719309.3 | 316 | Silent Mutation | TCA,TCG | S,S 25 | XP_006719372.1 |