Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACTGTACACATACGATATGTCC[A/G]TGGGGTTCTGTGAGATAATTAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605219 MIM: 610034 | ||||||||||||||||||||
Literature Links: |
DIABLO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DIABLO - diablo IAP-binding mitochondrial protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101593348 - uncharacterized LOC101593348 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS33A - VPS33A, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022916.4 | 1561 | Missense Mutation | ACG,ATG | T,M 483 | NP_075067.2 |