Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACTACTCTGCCTGCTCCAGCTT[C/G]AAACCAGGGTCCTACCACCCCACGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601190 | ||||||||||||||||||||
Literature Links: |
PDE6H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE6H - phosphodiesterase 6H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006205.2 | 384 | Nonsense Mutation | TCA,TGA | S,* 12 | NP_006196.1 | |
XM_017019431.1 | 384 | Nonsense Mutation | TCA,TGA | S,* 12 | XP_016874920.1 |