Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGGTGAGGGCGAAGTCTCCCGC[A/G]CAGCTGCAGTACACAGGCATGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600844 MIM: 174762 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
P2RX2 - purinergic receptor P2X 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLE - DNA polymerase epsilon, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006231.3 | 6923 | Silent Mutation | TGC,TGT | C,C 2238 | NP_006222.2 | |
XM_011534795.2 | 6923 | Intron | XP_011533097.1 | |||
XM_011534797.2 | 6923 | Intron | XP_011533099.1 | |||
XM_011534798.1 | 6923 | Intron | XP_011533100.1 | |||
XM_011534799.2 | 6923 | Intron | XP_011533101.1 | |||
XM_011534800.2 | 6923 | Intron | XP_011533102.1 | |||
XM_011534802.2 | 6923 | Intron | XP_011533104.1 |