Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCTGGTACGAGTGTGTGAAAAC[A/G]TCCCCATTGTGTTGTGTGGCAACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601179 | ||||||||||||||||||||
Literature Links: |
RAN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAN - RAN, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300796.1 | 432 | Missense Mutation | ATC,GTC | I,V 27 | NP_001287725.1 | |
NM_001300797.1 | 432 | Missense Mutation | ATC,GTC | I,V 27 | NP_001287726.1 | |
NM_006325.4 | 432 | Missense Mutation | ATC,GTC | I,V 115 | NP_006316.1 | |
XM_017019772.1 | 432 | Missense Mutation | ATC,GTC | I,V 115 | XP_016875261.1 | |
XM_017019773.1 | 432 | Missense Mutation | ATC,GTC | I,V 27 | XP_016875262.1 |