Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCAGCAGGCTGTCCCCCTCCAA[C/G]GCCTGGCTAATAATGTACTCTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SP1 - Sp1 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251825.1 | 671 | Missense Mutation | CGC,GGC | R,G 192 | NP_001238754.1 | |
NM_003109.1 | 671 | Missense Mutation | CGC,GGC | R,G 233 | NP_003100.1 | |
NM_138473.2 | 671 | Missense Mutation | CGC,GGC | R,G 240 | NP_612482.2 | |
XM_011538696.2 | 671 | Missense Mutation | CGC,GGC | R,G 233 | XP_011536998.1 |