Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTAAAACAGAACAGTCCACTTTG[A/G]AAAGTGAAGAATGGAATCCTTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM5 - transmembrane protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278237.1 | 361 | UTR 5 | NP_001265166.1 | |||
NM_014254.2 | 361 | Missense Mutation | AAA,GAA | K,E 62 | NP_055069.1 | |
XM_005268562.3 | 361 | UTR 5 | XP_005268619.1 | |||
XM_005268563.3 | 361 | UTR 5 | XP_005268620.1 | |||
XM_017018686.1 | 361 | UTR 5 | XP_016874175.1 | |||
XM_017018687.1 | 361 | UTR 5 | XP_016874176.1 |