Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGCCCCTCCTCCTGCAGGCGAA[A/C]GCAAGAAGATGACAGGGACGGTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120140 | ||||||||||||||||||||
Literature Links: |
COL2A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL2A1 - collagen type II alpha 1 chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM106C - transmembrane protein 106C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143841.1 | 236 | Missense Mutation | AAG,ACG | K,T 17 | NP_001137313.1 | |
NM_001143842.1 | 236 | Missense Mutation | AAG,ACG | K,T 17 | NP_001137314.1 | |
NM_001143843.1 | 236 | Missense Mutation | AAG,ACG | K,T 17 | NP_001137315.1 | |
NM_024056.3 | 236 | Missense Mutation | AAG,ACG | K,T 17 | NP_076961.1 |