Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGCATTGGTATGGAACCAAAG[A/T]TCCTCCATTCCTAATGATGAAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616381 | ||||||||||||||||||||
Literature Links: |
ZCCHC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZCCHC8 - zinc finger CCHC-type containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017612.4 | 1304 | Missense Mutation | AAC,ATC | N,I 384 | NP_060082.2 | |
XM_006719491.2 | 1304 | Missense Mutation | AAC,ATC | N,I 307 | XP_006719554.1 | |
XM_011538555.2 | 1304 | Missense Mutation | AAC,ATC | N,I 285 | XP_011536857.1 | |
XM_017019606.1 | 1304 | Missense Mutation | AAC,ATC | N,I 293 | XP_016875095.1 |