Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAAGAGGATATTCTTAGATTTC[C/T]GGATAGTGACTCTAGTAACTCCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601234 | ||||||||||||||||||||
Literature Links: |
NACA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NACA - nascent polypeptide-associated complex alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113201.2 | 572 | Missense Mutation | CAG,CGG | Q,R 97 | NP_001106672.1 | |
NM_001113202.1 | 572 | Missense Mutation | CAG,CGG | Q,R 97 | NP_001106673.1 | |
NM_001113203.2 | 572 | Missense Mutation | CAG,CGG | Q,R 807 | NP_001106674.2 | |
NM_001320193.1 | 572 | Intron | NP_001307122.1 | |||
NM_001320194.1 | 572 | Missense Mutation | CAG,CGG | Q,R 97 | NP_001307123.1 | |
NM_005594.5 | 572 | Missense Mutation | CAG,CGG | Q,R 97 | NP_005585.1 | |
XM_006719412.1 | 572 | Missense Mutation | CAG,CGG | Q,R 1960 | XP_006719475.1 | |
XM_006719413.2 | 572 | Missense Mutation | CAG,CGG | Q,R 1960 | XP_006719476.1 | |
XM_006719414.1 | 572 | Missense Mutation | CAG,CGG | Q,R 1960 | XP_006719477.1 | |
XM_006719418.1 | 572 | Intron | XP_006719481.1 | |||
XM_006719420.3 | 572 | Missense Mutation | CAG,CGG | Q,R 97 | XP_006719483.1 | |
XM_011538389.1 | 572 | Missense Mutation | CAG,CGG | Q,R 1960 | XP_011536691.1 | |
XM_017019337.1 | 572 | Intron | XP_016874826.1 |