Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTTATTCTTTTTAAGGTATTA[A/G]AAGAACTCATAATATATGTTTTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608368 | ||||||||||||||||||||
Literature Links: |
RAD9B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD9B - RAD9 checkpoint clamp component B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286531.1 | 361 | Missense Mutation | AAA,AGA | K,R 60 | NP_001273460.1 | |
NM_001286532.1 | 361 | Missense Mutation | AAA,AGA | K,R 60 | NP_001273461.1 | |
NM_001286533.1 | 361 | UTR 5 | NP_001273462.1 | |||
NM_001286534.1 | 361 | UTR 5 | NP_001273463.1 | |||
NM_001286535.1 | 361 | Missense Mutation | AAA,AGA | K,R 132 | NP_001273464.1 | |
NM_001286536.1 | 361 | Intron | NP_001273465.1 | |||
NM_152442.3 | 361 | Missense Mutation | AAA,AGA | K,R 132 | NP_689655.3 | |
XM_005253849.4 | 361 | Intron | XP_005253906.1 | |||
XM_011537971.2 | 361 | Missense Mutation | AAA,AGA | K,R 132 | XP_011536273.1 | |
XM_011537972.2 | 361 | Missense Mutation | AAA,AGA | K,R 108 | XP_011536274.1 | |
XM_011537974.2 | 361 | Missense Mutation | AAA,AGA | K,R 60 | XP_011536276.1 | |
XM_017018877.1 | 361 | Missense Mutation | AAA,AGA | K,R 80 | XP_016874366.1 | |
XM_017018878.1 | 361 | Missense Mutation | AAA,AGA | K,R 60 | XP_016874367.1 | |
XM_017018879.1 | 361 | Intron | XP_016874368.1 | |||
XM_017018880.1 | 361 | Intron | XP_016874369.1 | |||
XM_017018881.1 | 361 | Intron | XP_016874370.1 | |||
XM_017018882.1 | 361 | UTR 5 | XP_016874371.1 |