Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTCGTGAAGGACTGTGACCAGC[A/T]TAGAAAGGCTGCTCAGTGTGATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186711 MIM: 607081 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD27 - CD27 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242.4 | 396 | Missense Mutation | CAT,CTT | H,L 56 | NP_001233.1 | |
XM_011521042.2 | 396 | Intron | XP_011519344.1 | |||
XM_017020232.1 | 396 | Missense Mutation | CAT,CTT | H,L 56 | XP_016875721.1 | |
XM_017020233.1 | 396 | Intron | XP_016875722.1 | |||
XM_017020234.1 | 396 | Missense Mutation | CAT,CTT | H,L 56 | XP_016875723.1 |
CD27-AS1 - CD27 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAPBPL - TAP binding protein like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |