Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCCTGGTCATACTCCTCCTCAT[C/T]ATCATTTTTCTTCTCCTCCTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601657 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCER1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCER1 - coiled-coil glutamate rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152638.3 | 1290 | Missense Mutation | AAT,GAT | N,D 286 | NP_689851.1 |
EPYC - epiphycan | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LINC00615 - long intergenic non-protein coding RNA 615 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |