Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605476 MIM: 123829 MIM: 613336 MIM: 181035 | ||||||||||||||||||||
Literature Links: |
AGAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGAP2 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDK4 - cyclin dependent kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MARCH9 - membrane associated ring-CH-type finger 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6759 - microRNA 6759 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN31 - tetraspanin 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005981.3 | 1355 | Missense Mutation | ATC,ATG | I,M 92 | NP_005972.1 | |
XM_005269074.3 | 1355 | Missense Mutation | ATC,ATG | I,M 182 | XP_005269131.2 | |
XM_017019793.1 | 1355 | Intron | XP_016875282.1 |