Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGATGTTGTTTATCGAGGAGA[A/C]ATGCTTCTGGAGAAGATACAAAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603800 | ||||||||||||||||||||
Literature Links: |
MED21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED21 - mediator complex subunit 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271811.1 | 392 | UTR 3 | NP_001258740.1 | |||
NM_004264.4 | 392 | Missense Mutation | GAA,GAC | E,D 112 | NP_004255.2 |