Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACTTCACCTTCTCCGGGGAGGT[C/T]AACGTGGAGATCGCGTGCCGGAACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRHDE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRHDE - thyrotropin releasing hormone degrading enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013381.2 | 636 | Silent Mutation | GTC,GTT | V,V 167 | NP_037513.1 | |
XM_005268819.4 | 636 | Silent Mutation | GTC,GTT | V,V 212 | XP_005268876.1 | |
XM_011538248.2 | 636 | Intron | XP_011536550.1 | |||
XM_017019243.1 | 636 | Silent Mutation | GTC,GTT | V,V 212 | XP_016874732.1 | |
XM_017019244.1 | 636 | Intron | XP_016874733.1 |
TRHDE-AS1 - TRHDE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |