Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCTCTTTTGTGGTTTTGTTTT[C/T]GATGATGTTTCAGCAATACTGGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610142 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP290 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP290 - centrosomal protein 290 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMTC3 - transmembrane and tetratricopeptide repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181783.3 | 304 | Silent Mutation | TTC,TTT | F,F 30 | NP_861448.2 | |
XM_011537980.2 | 304 | Silent Mutation | TTC,TTT | F,F 30 | XP_011536282.1 | |
XM_011537981.2 | 304 | Intron | XP_011536283.1 |