Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCCAAAGTTGGCATCTTTGGA[C/T]ATCTCCACCTCCAGGCAGCGGCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606251 | ||||||||||||||||||||
Literature Links: |
GALNT9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT9 - polypeptide N-acetylgalactosaminyltransferase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122636.1 | 896 | Missense Mutation | ATA,ATG | I,M 572 | NP_001116108.1 | |
NM_021808.3 | 896 | Missense Mutation | ATA,ATG | I,M 206 | NP_068580.2 | |
XM_017019371.1 | 896 | Missense Mutation | ATA,ATG | I,M 471 | XP_016874860.1 | |
XM_017019372.1 | 896 | Missense Mutation | ATA,ATG | I,M 469 | XP_016874861.1 | |
XM_017019373.1 | 896 | Missense Mutation | ATA,ATG | I,M 469 | XP_016874862.1 | |
XM_017019374.1 | 896 | Missense Mutation | ATA,ATG | I,M 429 | XP_016874863.1 | |
XM_017019375.1 | 896 | Intron | XP_016874864.1 | |||
XM_017019376.1 | 896 | Intron | XP_016874865.1 |