Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAATCAGGTCCTCTGCATCAGA[A/G]AAGTTCAGCACTTGGTCCCCTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604527 MIM: 609504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNH3 - potassium voltage-gated channel subfamily H member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCRS1 - microspherule protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012300.1 | 1051 | Silent Mutation | TTC,TTT | F,F 294 | NP_001012300.1 | |
NM_001278341.1 | 1051 | Silent Mutation | TTC,TTT | F,F 90 | NP_001265270.1 | |
NM_006337.4 | 1051 | Silent Mutation | TTC,TTT | F,F 281 | NP_006328.2 | |
XM_005268572.3 | 1051 | Silent Mutation | TTC,TTT | F,F 281 | XP_005268629.1 | |
XM_011537760.2 | 1051 | Silent Mutation | TTC,TTT | F,F 332 | XP_011536062.1 | |
XM_017018689.1 | 1051 | Silent Mutation | TTC,TTT | F,F 294 | XP_016874178.1 | |
XM_017018690.1 | 1051 | Silent Mutation | TTC,TTT | F,F 294 | XP_016874179.1 |