Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTACCTTTTGATTTTTCCACTTG[A/T]GGAGTTTCATCCATTTCAACAAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615079 | ||||||||||||||||||||
Literature Links: |
ASUN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASUN - asunder, spermatogenesis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018164.2 | 1996 | Silent Mutation | CCA,CCT | P,P 642 | NP_060634.2 | |
XM_005253422.3 | 1996 | Silent Mutation | CCA,CCT | P,P 585 | XP_005253479.1 | |
XM_011520750.2 | 1996 | Silent Mutation | CCA,CCT | P,P 648 | XP_011519052.1 | |
XM_011520751.2 | 1996 | Silent Mutation | CCA,CCT | P,P 591 | XP_011519053.1 | |
XM_011520752.2 | 1996 | Silent Mutation | CCA,CCT | P,P 573 | XP_011519054.1 | |
XM_011520753.2 | 1996 | Silent Mutation | CCA,CCT | P,P 370 | XP_011519055.1 | |
XM_017019632.1 | 1996 | Silent Mutation | CCA,CCT | P,P 642 | XP_016875121.1 | |
XM_017019633.1 | 1996 | Silent Mutation | CCA,CCT | P,P 623 | XP_016875122.1 | |
XM_017019634.1 | 1996 | Silent Mutation | CCA,CCT | P,P 617 | XP_016875123.1 | |
XM_017019635.1 | 1996 | Silent Mutation | CCA,CCT | P,P 585 | XP_016875124.1 | |
XM_017019636.1 | 1996 | Silent Mutation | CCA,CCT | P,P 567 | XP_016875125.1 | |
XM_017019637.1 | 1996 | Silent Mutation | CCA,CCT | P,P 364 | XP_016875126.1 |