Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATGATGAGGGGCTTCCCCCCGC[A/G]GTTGTTCTCACCCACATCCAGGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605148 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT6 - polypeptide N-acetylgalactosaminyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007210.3 | 2085 | Missense Mutation | NP_009141.2 | |||
XM_005268607.1 | 2085 | Missense Mutation | XP_005268664.1 | |||
XM_006719214.2 | 2085 | Missense Mutation | XP_006719277.1 | |||
XM_011537819.2 | 2085 | Missense Mutation | XP_011536121.1 | |||
XM_011537822.2 | 2085 | Missense Mutation | XP_011536124.1 | |||
XM_017018744.1 | 2085 | Missense Mutation | XP_016874233.1 | |||
XM_017018745.1 | 2085 | Missense Mutation | XP_016874234.1 |