Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCCCACTGGGGTCCCAGACCAT[C/G]GAGTGAGCCTCTCCCCCAAGCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605378 MIM: 611366 MIM: 604899 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AAAS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AAAS - aladin WD repeat nucleoporin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173466.1 | 1298 | Silent Mutation | TCC,TCG | S,S 368 | NP_001166937.1 | |
NM_015665.5 | 1298 | Silent Mutation | TCC,TCG | S,S 401 | NP_056480.1 | |
XM_011538778.1 | 1298 | Silent Mutation | TCC,TCG | S,S 401 | XP_011537080.1 | |
XM_011538780.1 | 1298 | Silent Mutation | TCC,TCG | S,S 368 | XP_011537082.1 |
C12orf10 - chromosome 12 open reading frame 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFDN5 - prefoldin subunit 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |