Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTGCGCTACAGCCTGGCCGAC[C/G]AGGCCTCCATGGACAGCGGGGCACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605093 | ||||||||||||||||||||
Literature Links: |
SH2B3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SH2B3 - SH2B adaptor protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291424.1 | 668 | Intron | NP_001278353.1 | |||
NM_005475.2 | 668 | Missense Mutation | CAG,GAG | Q,E 208 | NP_005466.1 | |
XM_005253818.4 | 668 | Missense Mutation | CAG,GAG | Q,E 208 | XP_005253875.1 | |
XM_005253819.4 | 668 | Missense Mutation | CAG,GAG | Q,E 208 | XP_005253876.1 | |
XM_006719180.3 | 668 | Intron | XP_006719243.1 | |||
XM_011537719.2 | 668 | Missense Mutation | CAG,GAG | Q,E 208 | XP_011536021.1 | |
XM_011537720.2 | 668 | Missense Mutation | CAG,GAG | Q,E 208 | XP_011536022.1 | |
XM_011537721.2 | 668 | Intron | XP_011536023.1 |