Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGACTTCAGTTCAGGGAGGGGGA[C/G]GACTCGCGATGAATGACAGGGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607844 | ||||||||||||||||||||
Literature Links: |
LEMD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LEMD3 - LEM domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167614.1 | 969 | Missense Mutation | CGA,GGA | R,G 315 | NP_001161086.1 | |
NM_014319.4 | 969 | Missense Mutation | CGA,GGA | R,G 315 | NP_055134.2 |