Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAACTCTTGCTCTGAGCCATAGC[A/G]ATCTGCAGAGGAACACAAAGGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606720 MIM: 611477 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENDOU PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENDOU - endonuclease, poly(U) specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172439.1 | 913 | Missense Mutation | CGC,TGC | R,C 252 | NP_001165910.1 | |
NM_001172440.1 | 913 | Missense Mutation | CGC,TGC | R,C 189 | NP_001165911.1 | |
NM_006025.3 | 913 | Missense Mutation | CGC,TGC | R,C 211 | NP_006016.1 |
LOC105369748 - uncharacterized LOC105369748 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPAP3 - RNA polymerase II associated protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |