Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTATCGCCACACAGCCGCCAGTC[C/G]CTCACCAAACAGGGACTGGATACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FREM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FREM2 - FRAS1 related extracellular matrix protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207361.5 | 1127 | Missense Mutation | CCC,CGC | P,R 273 | NP_997244.4 | |
XM_017020554.1 | 1127 | Missense Mutation | CCC,CGC | P,R 273 | XP_016876043.1 |
LINC00437 - long intergenic non-protein coding RNA 437 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |