Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCTGCCGGCCACGATGCCGAAC[A/G]TGCTGCTGCCGCCCAAGGAGAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAA16 - N(alpha)-acetyltransferase 16, NatA auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110798.1 | 310 | Missense Mutation | ATG,GTG | M,V 4 | NP_001104268.1 | |
NM_018527.3 | 310 | Missense Mutation | ATG,GTG | M,V 4 | NP_060997.2 | |
NM_024561.4 | 310 | Missense Mutation | ATG,GTG | M,V 4 | NP_078837.3 | |
XM_005266523.2 | 310 | Intron | XP_005266580.1 | |||
XM_006719866.2 | 310 | Missense Mutation | ATG,GTG | M,V 4 | XP_006719929.1 | |
XM_011535227.2 | 310 | Missense Mutation | ATG,GTG | M,V 4 | XP_011533529.1 | |
XM_011535228.1 | 310 | Missense Mutation | ATG,GTG | M,V 4 | XP_011533530.1 | |
XM_017020744.1 | 310 | Intron | XP_016876233.1 | |||
XM_017020745.1 | 310 | Missense Mutation | ATG,GTG | M,V 4 | XP_016876234.1 | |
XM_017020746.1 | 310 | Missense Mutation | ATG,GTG | M,V 4 | XP_016876235.1 |