Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTAATTACTTGATAAAATGAAGT[G/T]CATCGCTGTGAACAATTGACCCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189969 | ||||||||||||||||||||
Literature Links: |
GTF2F2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTF2F2 - general transcription factor IIF subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004128.2 | 1165 | Intron | NP_004119.1 | |||
XM_011535052.2 | 1165 | Intron | XP_011533354.1 | |||
XM_011535053.2 | 1165 | Intron | XP_011533355.1 | |||
XM_017020550.1 | 1165 | Intron | XP_016876039.1 | |||
XM_017020551.1 | 1165 | Intron | XP_016876040.1 |
KCTD4 - potassium channel tetramerization domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198404.2 | 1165 | Missense Mutation | GAA,GCA | E,A 254 | NP_940686.2 |