Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCCCCTGCCTTCAAGGTGGAC[A/G]GCGTAGGCTTCCTAAAACACGACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL4A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL4A1 - collagen type IV alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303110.1 | 13002 | Intron | NP_001290039.1 | |||
NM_001845.5 | 13002 | Missense Mutation | CCG,CTG | P,L 1647 | NP_001836.3 | |
XM_011521048.2 | 13002 | Missense Mutation | CCG,CTG | P,L 1583 | XP_011519350.1 |