Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAAAGTACACCTTTCACAGCAG[C/T]CTTAAAGTTTGCAGCAGAAGAAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610553 | ||||||||||||||||||||
Literature Links: |
UFM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UFM1 - ubiquitin fold modifier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286703.1 | 238 | Missense Mutation | GCC,GTC | A,V 30 | NP_001273632.1 | |
NM_001286704.1 | 238 | Missense Mutation | GCC,GTC | A,V 50 | NP_001273633.1 | |
NM_001286705.1 | 238 | Missense Mutation | GCC,GTC | A,V 32 | NP_001273634.1 | |
NM_001286706.1 | 238 | Missense Mutation | GCC,GTC | A,V 32 | NP_001273635.1 | |
NM_016617.3 | 238 | Missense Mutation | GCC,GTC | A,V 32 | NP_057701.1 |