Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGCTTTCGAGGAGGAAGAAGAC[G/A]TCGCTCTGGTTTGTGGGGTCTCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC105377805 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105377805 - basic salivary proline-rich protein 4-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM255B - transmembrane protein 255B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182614.2 | 99 | Silent Mutation | ACA,ACG | T,T 24 | NP_872420.1 | |
XM_005272413.3 | 99 | Silent Mutation | ACA,ACG | T,T 24 | XP_005272470.1 | |
XM_005272415.2 | 99 | UTR 5 | XP_005272472.1 | |||
XM_006719986.2 | 99 | Intron | XP_006720049.1 | |||
XM_017020558.1 | 99 | Silent Mutation | ACA,ACG | T,T 24 | XP_016876047.1 | |
XM_017020559.1 | 99 | Intron | XP_016876048.1 | |||
XM_017020560.1 | 99 | Silent Mutation | ACA,ACG | T,T 24 | XP_016876049.1 |