Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATGGTATGAACTTTTCCAACTT[C/G]GCAACTGTACATTTCCCCATCTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608102 MIM: 605653 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLN5 - ceroid-lipofuscinosis, neuronal 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006493.2 | 599 | Missense Mutation | CGC,GGC | R,G 191 | NP_006484.1 | |
XM_011534917.2 | 599 | Missense Mutation | CGC,GGC | R,G 191 | XP_011533219.1 |
FBXL3 - F-box and leucine rich repeat protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012158.2 | 599 | Intron | NP_036290.1 | |||
XM_005266336.1 | 599 | Intron | XP_005266393.1 | |||
XM_005266337.2 | 599 | Intron | XP_005266394.1 | |||
XM_017020538.1 | 599 | Intron | XP_016876027.1 |