Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTAGATGTGGTCGTGCTCCCGAC[A/C]CCGCAGTGCCTCCTGGACCCGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602654 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FARP1 - FERM, ARH/RhoGEF and pleckstrin domain protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001715.3 | 449 | Intron | NP_001001715.2 | |||
NM_001286839.1 | 449 | Intron | NP_001273768.1 | |||
NM_005766.3 | 449 | Intron | NP_005757.1 | |||
XM_011521046.2 | 449 | Intron | XP_011519348.1 | |||
XM_017020312.1 | 449 | Intron | XP_016875801.1 | |||
XM_017020313.1 | 449 | Intron | XP_016875802.1 |
RNF113B - ring finger protein 113B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178861.4 | 449 | Missense Mutation | GGT,TGT | G,C 140 | NP_849192.1 |