Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTTTGGCGACAGTGAGGAGGA[C/T]GGAGAGGATGTGTTCGAGGTGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611626 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPHOSPH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPHOSPH8 - M-phase phosphoprotein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017520.3 | 263 | Silent Mutation | GAC,GAT | D,D 54 | NP_059990.2 | |
XM_011535124.2 | 263 | Silent Mutation | GAC,GAT | D,D 54 | XP_011533426.1 |