Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGCGGCAGGTCAGCCCCGAAGC[A/G]CTGGAAGGGAAGATCCTGGCTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BIVM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BIVM - basic, immunoglobulin-like variable motif containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BIVM-ERCC5 - BIVM-ERCC5 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204425.1 | 489 | Intron | NP_001191354.1 |
ERCC5 - ERCC excision repair 5, endonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000123.3 | 489 | Silent Mutation | GCA,GCG | A,A 21 | NP_000114.2 |