Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTCATCCTCTTCATCTTCCTC[A/C]TCTTCCTCCTCTTCCTTCTTTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 163905 | ||||||||||||||||||||
Literature Links: |
HMGB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HMGB1 - high mobility group box 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313892.1 | 1511 | Missense Mutation | GAG,GAT | E,D 190 | NP_001300821.1 | |
NM_001313893.1 | 1511 | Missense Mutation | GAG,GAT | E,D 190 | NP_001300822.1 | |
NM_002128.5 | 1511 | Missense Mutation | GAG,GAT | E,D 190 | NP_002119.1 |