Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCACGCGAGCCTCCTCGGTGATA[A/C]CAGCCACTTGGGTTTACCAGGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN10 - claudin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160100.1 | 350 | Missense Mutation | ACA,CCA | T,P 39 | NP_001153572.1 | |
NM_006984.4 | 350 | Intron | NP_008915.1 | |||
NM_182848.3 | 350 | Missense Mutation | ACA,CCA | T,P 39 | NP_878268.1 | |
XM_011521134.2 | 350 | Intron | XP_011519436.1 | |||
XM_017020843.1 | 350 | Missense Mutation | ACA,CCA | T,P 39 | XP_016876332.1 | |
XM_017020844.1 | 350 | Missense Mutation | ACA,CCA | T,P 39 | XP_016876333.1 |