Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTTTTGGATTTTCACTGAGAA[C/T]GACCTTCCCTTAATCGTCTTCTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608777 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POSTN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POSTN - periostin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135934.1 | 2454 | Missense Mutation | CAT,CGT | H,R 777 | NP_001129406.1 | |
NM_001135935.1 | 2454 | Missense Mutation | CAT,CGT | H,R 779 | NP_001129407.1 | |
NM_001135936.1 | 2454 | Missense Mutation | CAT,CGT | H,R 749 | NP_001129408.1 | |
NM_001286665.1 | 2454 | Missense Mutation | CAT,CGT | H,R 807 | NP_001273594.1 | |
NM_001286666.1 | 2454 | Missense Mutation | CAT,CGT | H,R 747 | NP_001273595.1 | |
NM_001286667.1 | 2454 | Missense Mutation | CAT,CGT | H,R 719 | NP_001273596.1 | |
NM_006475.2 | 2454 | Missense Mutation | CAT,CGT | H,R 834 | NP_006466.2 | |
XM_005266231.2 | 2454 | Missense Mutation | CAT,CGT | H,R 806 | XP_005266288.1 | |
XM_005266232.2 | 2454 | Missense Mutation | CAT,CGT | H,R 804 | XP_005266289.1 | |
XM_017020355.1 | 2454 | Intron | XP_016875844.1 | |||
XM_017020356.1 | 2454 | Intron | XP_016875845.1 |