Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGCCGTTGCCATTATTATGAATC[A/G]AATTTTGCTTTCTTTGACAATGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610351 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP4R3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP4R3A - protein phosphatase 4 regulatory subunit 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284280.1 | 2576 | Silent Mutation | TTC,TTT | F,F 818 | NP_001271209.1 | |
NM_001284281.1 | 2576 | Silent Mutation | TTC,TTT | F,F 592 | NP_001271210.1 | |
XM_005267842.2 | 2576 | Silent Mutation | TTC,TTT | F,F 831 | XP_005267899.1 |