Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATAGGACACCCACCTTGTCCCC[A/G]GCCAGGATCTTGAAGGAAGCCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102575 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTN1 - actinin alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102.3 | 3463 | Silent Mutation | GCC,GCT | A,A 837 | NP_001093.1 | |
NM_001130004.1 | 3463 | Silent Mutation | GCC,GCT | A,A 859 | NP_001123476.1 | |
NM_001130005.1 | 3463 | Silent Mutation | GCC,GCT | A,A 832 | NP_001123477.1 | |
XM_011537265.2 | 3463 | Silent Mutation | GCC,GCT | A,A 874 | XP_011535567.1 | |
XM_011537266.2 | 3463 | Silent Mutation | GCC,GCT | A,A 869 | XP_011535568.1 | |
XM_011537267.2 | 3463 | Silent Mutation | GCC,GCT | A,A 866 | XP_011535569.1 | |
XM_011537268.2 | 3463 | Silent Mutation | GCC,GCT | A,A 861 | XP_011535570.1 | |
XM_017021720.1 | 3463 | Silent Mutation | GCC,GCT | A,A 1051 | XP_016877209.1 | |
XM_017021721.1 | 3463 | Silent Mutation | GCC,GCT | A,A 1043 | XP_016877210.1 | |
XM_017021722.1 | 3463 | Silent Mutation | GCC,GCT | A,A 1022 | XP_016877211.1 | |
XM_017021723.1 | 3463 | Silent Mutation | GCC,GCT | A,A 1014 | XP_016877212.1 | |
XM_017021725.1 | 3463 | Silent Mutation | GCC,GCT | A,A 1001 | XP_016877214.1 | |
XM_017021726.1 | 3463 | Silent Mutation | GCC,GCT | A,A 993 | XP_016877215.1 | |
XM_017021727.1 | 3463 | Silent Mutation | GCC,GCT | A,A 840 | XP_016877216.1 | |
XM_017021728.1 | 3463 | Silent Mutation | GCC,GCT | A,A 811 | XP_016877217.1 |