Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGCCCCAGCATAGTTGGCAAAC[A/T]GGGTGCTGAGCAGCTTGAGGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613613 MIM: 160760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR208B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR208B - microRNA 208b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYH7 - myosin, heavy chain 7, cardiac muscle, beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000257.3 | 1925 | Missense Mutation | CAG,CTG | Q,L 620 | NP_000248.2 | |
XM_017021340.1 | 1925 | Missense Mutation | CAG,CTG | Q,L 620 | XP_016876829.1 |